Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01257

Millipore

Starch

soluble acc. to Zulkowsky GR for analysis

Synonym(s):

Starch, Amylum, Potato starch for determination of diastase

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
MDL number:
UNSPSC Code:
12352201
EC Index Number:
232-686-4

Quality Level

form

solid

pH

6.0-7.5 (25 °C, 20 g/L in H2O)

solubility

50 g/L

bulk density

300 kg/m3

storage temp.

no temp limit

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11+,12-/m1/s1

InChI key

GUBGYTABKSRVRQ-ASMJPISFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carbohydrate
This starch quality is prepared according to a modified procedure published by Zulkowsky. The molecular weight is reduced to a mean by hydrolysis and oxidation.

Analysis Note

Appearance (colour): white
Appearance (description): fine powder
pH-value (2 %; water): 6.5 - 7.5
Sensitivity: passes test
Reducing matter (as maltose): ≤ 1.0 %
Sulfated ash: ≤ 7.0 %
Loss on drying (105 °C, 2 h): ≤ 10.0 %
Suitability as enzyme substrate (for amylases): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service