Skip to Content
MilliporeSigma

1.01131

SAFC

Ammonium hydrogen carbonate

EMPROVE® ESSENTIAL, Ph. Eur., BP, E 503

Pharma Manufacturing

Synonym(s):

Ammonium hydrogen carbonate, Ammonium bicarbonate, Ammonium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)HCO3
CAS Number:
Molecular Weight:
79.06
MDL number:
UNSPSC Code:
12352301
E Number:
E503
EC Index Number:
213-911-5

Agency

BP
Ph. Eur.

Quality Level

vapor pressure

67 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

Assay

99.0-101.0% (acidimetric)

form

solid

potency

1576 mg/kg LD50, oral (Rat)

pH

8 (20 °C, 50 g/L in H2O)

mp

106 °C

application(s)

pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[Zn+2].[Zn+2].N.N.[O-]C(=O)[O-].[O-]C(=O)[O-].OC(=O)O

InChI

1S/3CH2O3.2H3N.2Zn/c3*2-1(3)4;;;;/h3*(H2,2,3,4);2*1H3;;/q;;;;;2*+2/p-4

InChI key

QOPSSYZLVIENBR-UHFFFAOYSA-J

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ammonium hydrogen carbonate is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service