Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

1.00964

Supelco

Tetrachloroethylene

EMPLURA®

Synonym(s):

Tetrachloroethylene, Ethylene tetrachloride, Perchloroethylene, PCE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2Cl4
CAS Number:
Molecular Weight:
165.83
MDL number:
UNSPSC Code:
12191502
EC Index Number:
204-825-9
NACRES:
NA.05

vapor pressure

19 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

potency

2629 mg/kg LD50, oral (Rat)

impurities

≤0.001% Free acid (as HCl)
≤0.005% Water

evapn. residue

≤0.001%

bp

121 °C/1013 hPa

mp

-22 °C

solubility

0.16 g/L

density

1.62 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Certified reference samples of tetrachloromethane, tetrachloroethylene, and chlorophenol solutions.: This study presents certified reference samples for tetrachloromethane, tetrachloroethylene, and chlorophenol solutions, crucial for standardizing measurements in analytical chemistry (Piterskikh et al., 2000).

  • Continuous Determination of High-Vapor-Phase Concentrations of Tetrachloroethylene Using On-Line Mass Spectrometry.: This research focuses on the continuous monitoring of high vapor-phase concentrations of tetrachloroethylene using on-line mass spectrometry, improving real-time detection capabilities (Fine et al., 2008).

  • An Analytical Solution of Tetrachloroethylene Transport and Biodegradation.: The paper provides an analytical solution for the transport and biodegradation of tetrachloroethylene in porous media, essential for environmental remediation strategies (Sun et al., 2004).

  • Thermal decomposition of tetrachloroethylene with excess hydrogen.: This study examines the thermal decomposition of tetrachloroethylene in the presence of excess hydrogen, contributing to understanding its breakdown products and potential applications in chemical engineering (Won, 2009).

  • A Practical Synthesis of Tetrachloroethylene Oxide.: This article discusses a practical synthesis method for tetrachloroethylene oxide, an important intermediate in chemical synthesis and industrial applications (Campbell & Vogl, 1977).

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as HCl): ≤ 0.001 %
Density (d 20 °C/ 4 °C): 1.621 - 1.623
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service