Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00307

Supelco

Hydrobromic acid

47%, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Hydrogen bromide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

Product Name

Hydrobromic acid 47%, for analysis EMSURE® ACS,ISO

grade

ACS reagent

Quality Level

Agency

reag. ISO

vapor pressure

10.6 hPa ( 20 °C)

product line

EMSURE®

form

liquid

concentration

47.0-49.0% HBr

impurities

≤0.01 ppm Se (Selenium)
≤0.02 ppm Be (Beryllium)
≤0.02 ppm Co (Cobalt)
≤0.02 ppm Cu (Copper)
≤0.02 ppm Li (Lithium)
≤0.02 ppm Ni (Nickel)
≤0.02 ppm Pb (Lead)
≤0.02 ppm Sr (Strontium)
≤0.05 ppm Cd (Cadmium)
≤0.05 ppm Ge (Germanium)
≤0.05 ppm Mn (Manganese)
≤0.05 ppm Mo (Molybdenum)
≤0.05 ppm Tl (Thallium)
≤0.05 ppm V (Vanadium)
≤0.1 ppm Al (Aluminium)
≤0.1 ppm As (Arsenic)
≤0.1 ppm Ba (Barium)
≤0.1 ppm Bi (Bismuth)
≤0.1 ppm Cr (Chromium)
≤0.1 ppm Fe (Iron)
≤0.1 ppm K (Potassium)
≤0.1 ppm Mg (Magnesium)
≤0.1 ppm Ti (Titanium)
≤0.1 ppm Zn (Zinc)
≤0.1 ppm Zr (Zirconium)
≤0.5 ppm Ca (Calcium)
≤2 ppm Phosphate, Phosphite (as PO4)
≤20 ppm Iodide (I)
≤200 ppm Chloride (Cl)
≤30 ppm Sulfate and sulphite (as SO4)
≤5 ppm Heavy metals (ACS)
≤5 ppm Heavy metals (as Pb)

ign. residue

≤20 ppm (as sulfate)

bp

126 °C/1013 hPa

mp

-11 °C

density

1.49 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): 47.0 - 49.0 %
Chloride (Cl): ≤ 200 ppm
Iodide (I): ≤ 20 ppm
Phosphate, Phosphite (as PO₄): ≤ 2 ppm
Heavy metals (as Pb): ≤ 5 ppm
Heavy metals (ACS): ≤ 5 ppm
Sulfate and sulphite (as SO₄): ≤ 30 ppm
Al (Aluminium): ≤ 0.1 ppm
As (Arsenic): ≤ 0.1 ppm
Ba (Barium): ≤ 0.1 ppm
Be (Beryllium): ≤ 0.02 ppm
Bi (Bismuth): ≤ 0.1 ppm
Ca (Calcium): ≤ 0.5 ppm
Cd (Cadmium): ≤ 0.05 ppm
Co (Cobalt): ≤ 0.02 ppm
Cr (Chromium): ≤ 0.1 ppm
Cu (Copper): ≤ 0.02 ppm
Fe (Iron): ≤ 0.1 ppm
Ge (Germanium): ≤ 0.05 ppm
K (Potassium): ≤ 0.1 ppm
Li (Lithium): ≤ 0.02 ppm
Mg (Magnesium): ≤ 0.1 ppm
Mn (Manganese): ≤ 0.05 ppm
Mo (Molybdenum): ≤ 0.05 ppm
Ni (Nickel): ≤ 0.02 ppm
Pb (Lead): ≤ 0.02 ppm
Se (Selenium): ≤ 0.01 ppm
Sr (Strontium): ≤ 0.02 ppm
Ti (Titanium): ≤ 0.1 ppm
Tl (Thallium): ≤ 0.05 ppm
V (Vanadium): ≤ 0.05 ppm
Zn (Zinc): ≤ 0.1 ppm
Zr (Zirconium): ≤ 0.1 ppm
Residue on ignition (as sulfate): ≤ 20 ppm
Conforms to ACS, ISO

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service