Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18856

Sigma-Aldrich

Hydrogen bromide

(32% solution in acetic acid) for synthesis

Synonym(s):

HBr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.22

grade

for synthesis

Quality Level

vapor pressure

360 hPa ( 20 °C)

form

liquid

bp

<200 °C/1013 hPa

transition temp

flash point 39 °C (refers to pure substance)

density

1.38 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (argentometric): 30.0 - 34.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup - (refers to pure substance)

Flash Point(C)

39 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service