Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00266

Supelco

Colour Reference Solutions BY

for examination of the colour of solutions related to Ph. Eur. method 1 (BY1-BY7), Certipur®

Synonym(s):

Brownish yellow color reference standards (identified sequentially from BY1 to BY7)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

according to Ph. Eur. color intensity testing BY1-BY7

product line

Certipur®

application(s)

coatings
environmental
food and beverages
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

General description

The kit contains a set of seven brownish yellow color reference standards (identified sequentially from BY1 to BY7) and 3 empty sample cells. The solutions are produced from 3 primary metal salt solutions of ferric(III) chloride, cobalt(II) chloride, and copper(II) sulfate, in accordance with chapter 2.2.2 ′Degree of colouration of liquids′ of the European Pharmacopoeia (Ph. Eur.).

Application

The color reference standards are used as calibration and/or quality control standards to measure the degree of coloration of solutions according to Ph. Eur. Method 1.

Features and Benefits

  • Ready-to-use solutions
  • Prepared and qualified in accordance with Ph. Eur.
  • Stable solutions with a shelf life of 3 years
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Met. Corr. 1 - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service