Skip to Content
MilliporeSigma
All Photos(1)

Documents

83952

Supelco

Color Reference Solutions

related to Ph.Eur Color intensity testing, analytical standard, Set of colors B, BY, Y, GY, R

Synonym(s):

Color Calibration Solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

according to Ph. Eur.

shelf life

limited shelf life, expiry date on the label

packaging

ampule of 2 mL

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

The kit contains set of brown(83951), yellow (83883), red (87448), greenish-yellow (82995), and brownish-yellow (86293) color reference standards. The solutions are produced from three primary metal salt solutions of ferric(III) chloride, cobalt(II) chloride, and copper(II) sulfate pentahydrate, in accordance with chapter 2.2.2 ′Degree of colouration of liquids′ of the European Pharmacopoeia (Ph. Eur.).

Application

They are used as calibration and/or quality control standards to measure the degree of coloration of liquids in the brown-yellow-red color range.

Features and Benefits

  • Ready-to-use solutions
  • Prepared and qualified in accordance with Ph. Eur.
  • Available in tamper-evident 2 mL ampules
  • Expiry dated on the label with a limited shelf life
  • Accompanied with a detailed certificate of analysis for an effective quality management system

Components

This is a set of the following color reference solutions sets according to PhEur:

  • 83951-1SET-FColor Reference Solutions B acc. to PhEur - 10 X 2mL ampules
  • 86293-1SET-FColor Reference Solutions BY acc. to PhEur - 8 X 2mL ampules
  • 83883-1SET-FColor Reference Solutions Y acc. to PhEur - 8 X 2mL ampules
  • 82995-1SET-FColor Reference Solutions GY acc. to PhEur - 8 X 2mL ampules
  • 87448-1SET-FColor Reference Solutions R acc. to PhEur - 8 X 2mL ampules

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Met. Corr. 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service