Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

07-947

Sigma-Aldrich

Anti-dimethyl-Histone H4 (Arg3) Symmetric Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

histone 4, H4, histone H4, histone cluster 4, H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Arg3)

Gene Information

human ... H4C1(8359)
mouse ... H4C1(326619)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications, which include acetylation, phosphorylation, methylation, and ubiquitin-ation, occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Arginine modifications of histones proteins are mediated by two classes of protein arginine methyltransferases (PRMTs), class I and II. Class I PRMTs place two methyl groups on the one nitrogen atom of the arginine guanidino group, resulting in an asymmetric modification. In contrast class II PRMTs place two methyl groups on two nitrogen atoms of the guanidino group producing a symmetric dimethylation. Although there are exceptions, asymmetric modifications are typically associated with transcription activation while symmetric modifications are most commonly associated with transcriptional repression.

Specificity

Broad species cross-reactivity expected based on sequence similarity.
This antibody recognizes Histone H4 symmetrically dimethylated at Arg3.

Immunogen

Epitope: Symmetrically dimethylated Arg3
KLH-conjugated linear peptide corresponding to human Histone H4 when symmetrically dimethylated at Arg3.

Application

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Histone H4 in HeLa and A431 cells.

Peptide Dot Blot Analysis: 0.5 µg/mL of this antibody detected Histone H4 dimethyl Arg3 (symmetric) peptides, but did not detect Histone H4 dimethyl Arg3 (assymmetric) peptide, other modified peptides, or non-methyated histone H4 peptides.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-dimethyl-Histone H4 (Arg3) Symmetric Antibody (rabbit polyclonal antibody) validated in WB, ICC, DB to detect dimethyl-Histone H4 (Arg3) Symmetric also known as histone H4, histone cluster 4 H4.

Quality

Evaluated by Western Blot in HeLa acid extract.

Western Blot Analysis: 0.1 µg/mL of this antibody detected Histone H4 on 10 µg of HeLa acid extract.

Target description

~13 kDa observed. Crossreacting band at ~58 kDa has been observed in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of the arginine methyltransferase PRMT5-MEP50 reveals a mechanism for substrate specificity.
Ho, MC; Wilczek, C; Bonanno, JB; Xing, L; Seznec, J; Matsui, T; Carter, LG; Onikubo et al.
Testing null
Amuza Byaruhanga Lucky et al.
Communications biology, 6(1), 659-659 (2023-06-23)
Protein arginine methyltransferases (PRMTs) regulate many important cellular processes, such as transcription and RNA processing in model organisms but their functions in human malaria parasites are not elucidated. Here, we characterize PfPRMT5 in Plasmodium falciparum, which catalyzes symmetric dimethylation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service