Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-852

Sigma-Aldrich

Anti-phospho-MEK1 (Thr292) Antibody

Upstate®, from rabbit

Synonym(s):

MAP Kinase Kinase 1, MAPK/ERK Kinase 1, MAPKK1, MAP2K1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr292)

Gene Information

human ... MAP2K1(5604)

Specificity

Recognizes phospho-MEK1 (Thr292).

Immunogen

Peptide corresponding to amino acid region encompassing the human, mouse, rat and hamster phospho-MEK1 (Thr292).

Application

Anti-phospho-MEK1 (Thr292) Antibody is an antibody against phospho-MEK1 (Thr292) for use in WB.
Immunoblot Analysis: A 1:1000 dilution of this lot detected phosphorylated MEK1 (Thr292) in RIPA lysates of untreated A431 cells.

Quality

Routinely evaluated by immunoblot.

Target description

~44 kDa

Linkage

Replaces: 07-348

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual role of PID1 in regulating apoptosis induced by distinct anticancer-agents through AKT/Raf-1-dependent pathway in hepatocellular carcinoma.
Yang, et al.
Cell death discovery, 9, 139-139 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service