Skip to Content
MilliporeSigma

LSKMAGG

PureProteome Protein G Magnetic Bead System

The PureProteome Protein G Magnetic Bead System is a powerful system that helps researchers purify proteins by maximizing recovery and eliminating variability

Synonym(s):

Protein G Magnetic Beads, PureProteome Magnetic Beads

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41105507
NACRES:
NA.56
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 2 × 1 mL

manufacturer/tradename

PureProteome

technique(s)

depletion: suitable (serum), immunoprecipitation (IP): suitable, protein purification: suitable

particle size

10 μm

capacity

2.5-3.5 μg/μL, suspension binding capacity (rabbit IgG)

shipped in

wet ice

General description

The PureProteome Protein G Magnetic Beads contain Protein G, a binding protein for antibodies, which is coated on the surface of the beads. These beads are designed to isolate immunoglobulins (IgG) from complex mixtures through antibody-affinity purification. These beads are used in conjunction with a magnetic stand to immobilize the beads. Following this step, the sample is washed to remove any impurities, and then the immobilized IgG can be eluted for further analysis or experimentation.

Application

PureProteome Protein G Magnetic Bead System is suitable for:
  • serum depletion
  • immunoprecipitation
  • protein purification    

PureProteome Protein G Magnetic Bead System is suitable:

  • for isolation of the immunoglobulin G (IgG) fraction from a small volume of plasma
  • for co-immunoprecipitation (Co-IP)

Features and Benefits

  • PureProteome Magnetic Beads are very well suited for fully automated purification process using the KingFisher Duo particle processor, it is not dependent on the cell lysate clarification step
  • The entire process is reproducible, and the results are comparable to that of a standard protocol using the PureProteome Magnetic Stand
  • PureProteome Magnetic Beads is compatible with any automated system for low, medium, or high-throughput sample preparation.

Legal Information

PureProteome is a trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library