Skip to Content
MilliporeSigma
All Photos(2)

Documents

07-502-I

Sigma-Aldrich

Anti-p67-phox (not Human) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Neutrophil cytosol factor 2, NCF-2, 67 kDa neutrophil oxidase factor, NADPH oxidase activator 2, Neutrophil NADPH oxidase factor 2, p67-phox, p67-phox (rodent)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ncf2(17970)
rat ... Ncf2(364018)

Related Categories

General description

Neutrophil cytosol factor 2 (UniProt Q09014; also known as 67 kDa neutrophil oxidase factor, NADPH oxidase activator 2, NADPH oxidase subunit 67 kDa, NCF-2, Ncf2, Neutrophil NADPH oxidase factor 2, p67-phox, p47phox) is encoded by the murine Ncf2 (also known as Noxa2, p67phox) gene in murine species (Gene ID 17970). Activation of NADPH-oxidase dependent superoxide generation requires the participation of several cytosolic factors, including p47-phox, p67-phox, p40-phox and Rac. In the resting cell p47phox, p67phox, and p40phox form a large molecular weight cytosolic complex. Activation of the NADPH oxidase is initiated by the assembly of p47phox, p67phox, and Rac with cytochrome b558 in a 1:1:1:1 complex in the plasma membrane.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of mouse p67-phox.

Application

Anti-p67-phox (not Human) Antibody is an antibody against p67-phox, tested in mouse and rat, but is not expected to react with human, for use in Western Blotting and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected p67-phox (rodent) in rat colon and mouse hindbrain tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in C6 and THP-1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected p67-phox (rodent) in 10 µg of C6 and THP-1 cell lysate.

Target description

~72 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service