Skip to Content
MilliporeSigma
All Photos(4)

Documents

HPA009647

Sigma-Aldrich

Anti-LDLR antibody produced in rabbit

Ab1, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LDLCQ2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RTTLSNGGCQYLCLPAPQINPHSPKFTCACPDGMLLARDMRSCLTEAEAAVATQETSTVRLKVSSTAVRTQHTTTRPVPDTSRLPGATPGLTTVEIVTMSHQALGDVAGRGNEKKPSSV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LDLR(3949)

Looking for similar products? Visit Product Comparison Guide

General description

LDLR (low density lipoprotein receptor) is a transmembrane protein, which is composed of five structural domains- a ligand binding domain (LBD) in the N-terminal, an EGF (extracellular growth factor)-like domain, a small glycosylated domain, a transmembrane domain and a cytoplasmic domain present in the C-terminal, which is involved in localization and endocytosis of the LDLR.

Immunogen

Low-density lipoprotein receptor precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In liver and peripheral tissues, LDLR (low density lipoprotein receptor) plays a predominant role in the internalization of LDL and VLDL (very LDL) remnants. It internalizes circulating LDL and VLDL in endosomes, which is then degraded by lysosomes, whereas the receptor is recycled to cell surface. Polymorphisms in this gene are linked with familial hypercholesterolaemia (FH), which is an autosomal dominant disease, where the patients have elevated levels of low-density lipoprotein cholesterol (LDL-C).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71915

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Merel L Hartgers et al.
Current cardiology reports, 17(12), 109-109 (2015-10-21)
Familial hypercholesterolemia (FH) is an autosomal dominant genetic disorder that clinically leads to increased low density lipoprotein-cholesterol (LDL-C) levels. As a consequence, FH patients are at high risk for cardiovascular disease (CVD). Mutations are found in genes coding for the
Amna Khamis et al.
European journal of human genetics : EJHG, 23(6), 790-795 (2014-09-25)
Familial hypercholesterolaemia (FH) is an autosomal dominant inherited disease characterised by increased low-density lipoprotein cholesterol (LDL-C) levels. The functionality of four novel variants within the LDLR 5'UTR and promoter located at c.-13A>G, c.-101T>C, c.-121T>C and c.-215A>G was investigated using in
Juan Martínez-Oliván et al.
FEBS letters, 589(23), 3534-3540 (2015-11-04)
The LDL receptor (LDLR) internalizes LDL and VLDL particles. In the endosomes, it adopts a closed conformation important for recycling, by interaction of two modules of the ligand binding domain (LR4-5) and a β-propeller motif. Here, we investigate by SPR
R de Neergaard et al.
Andrology, 6(6), 817-828 (2018-09-06)
Cholesterol is essential for cell membrane stability, permeability, and fluidity. Cholesterol is present in seminal plasma, but whether a relationship between the level of cholesterol in seminal plasma and semen quality exists remains to be elucidated. To explore the association

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service