Skip to Content
MilliporeSigma
All Photos(1)

Documents

07-1375

Sigma-Aldrich

Anti-phospho-Androgen Receptor (Ser81) Antibody

Upstate®, from rabbit

Synonym(s):

Anti-AIS, Anti-AR8, Anti-DHTR, Anti-HUMARA, Anti-HYSP1, Anti-KD, Anti-NR3C4, Anti-SBMA, Anti-SMAX1, Anti-TFM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

dog (expected based only 1 amino acid difference), horse (expected based only 1 amino acid difference), primate (expected based only 1 amino acid difference), bovine (expected based only 1 amino acid difference)

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer81)

Gene Information

bovine ... Ar(280675)
dog ... Ar(403588)
human ... AR(367)

General description

The X-linked androgen receptor (Dihydrotestosterone receptor; Nuclear receptor subfamily 3 group C member 4) is a member of the family of NR3 ligand-dependent transcription factors characterized by a ligand binding domain at the C-terminus, a central DNA-binding domain, and N-terminal transcription- activation domains. Androgen receptors are activated by steroid hormones such as testosterone and dihydrotestosterone. Activated androgen receptors translocate from the cytoplasm to the nucleus where they form homodimers that bind DNA. The androgen receptor associates with a number of adaptor-like proteins and functions as a complex that may induce or repress transcription depending on whether its associated proteins are co-activators or co-repressors that affect transcription possibly by modifying chromatin; co-activators are involved in histone acetylation and transcriptional activation whereas co-repressors are involved in histone deactylation and transcriptional repression. The androgen receptor is important for determining sex and male characteristics. Defective androgen receptors may play a role in Reifenstein syndrome, Kennedy disease, as well as prostate cancer.

Specificity

Detects Androgen Receptor (AR) only when phosphorylated on Ser81

Immunogen

KLH-conjugated synthetic peptide corresponding to amino acids including and surrounding Ser81 of human Androgen Receptor.

Application

Anti-phospho-Androgen Receptor (Ser81) Antibody is an antibody against phospho-Androgen Receptor (Ser81) for use in WB.
Research Category
Signaling

Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Routinely evaluated by western blot on R1881 (methyltrienolone) treated LNCap lysates

Target description

110 kDa

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
R1881 (methyltrienolone) treated LNCap lysates

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Hormaechea-Agulla et al.
Molecular cancer, 16(1), 146-146 (2017-08-31)
The Ghrelin-system is a complex, pleiotropic family composed of several peptides, including native-ghrelin and its In1-ghrelin splicing variant, and receptors (GHSR 1a/b), which are dysregulated in various endocrine-related tumors, where they associate to pathophysiological features, but the presence, functional role
Nooshin Mirkheshti et al.
Oncotarget, 7(38), 62240-62254 (2016-08-25)
Androgen receptor (AR) and PI3K/AKT/mTORC1 are major survival signals that drive prostate cancer to a lethal disease. Reciprocal activation of these oncogenic pathways from negative cross talks contributes to low/limited success of pathway-selective inhibitors in curbing prostate cancer progression. We
J M Willder et al.
British journal of cancer, 108(1), 139-148 (2013-01-17)
Prostate cancer cell growth is dependent upon androgen receptor (AR) activation, which is regulated by specific kinases. The aim of the current study is to establish if AR phosphorylation by Cdk1 or ERK1/2 is of prognostic significance. Scansite 2.0 was
Milly J McAllister et al.
Prostate cancer and prostatic diseases, 23(4), 596-606 (2020-05-03)
Despite increases in diagnostics and effective treatments, over 300,000 men die from prostate cancer highlighting the need for specific and differentiating biomarkers. AR phosphorylation associates with castrate-resistance, with pARser213 promoting transcriptional activity. We hypothesise that combined pARser81 and pARser213 reduces
Kazumasa Komura et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(22), 6259-6264 (2016-05-18)
The androgen receptor (AR) plays an essential role in prostate cancer, and suppression of its signaling with androgen deprivation therapy (ADT) has been the mainstay of treatment for metastatic hormone-sensitive prostate cancer for more than 70 y. Chemotherapy has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service