Skip to Content

1086301

USP

Calcitriol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1α,25-Dihydroxycholecalciferol monohydrate, (1α,3β,5Z,7E)-9,10-Secocholesta-5,7,10(19)-triene-1,3,25-triol monohydrate, 1α,25-Dihydroxyvitamin D3 monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H44O3 · H2O
CAS Number:
Molecular Weight:
434.65
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

calcitriol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)
pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

O[C@H]1C[C@@H](C(=C)\C(=C/C=C2/[C@H]3[C@@]([C@H](CC3)[C@@H](CCCC(O)(C)C)C)(CCC/2)C)\C1)O.O

InChI

1S/C27H44O3.H2O/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2;/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3;1H2/b20-10+,21-11-;/t18-,22-,23-,24+,25+,27-;/m1./s1

InChI key

VMBWEMQRTFKRSU-RFHBVEJWSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calcitriol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Repr. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library