Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

06-1049

Sigma-Aldrich

Anti-Axin-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Axin-1, Axis inhibition protein 1, hAxin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Axin1(12005)

General description

Axin-1 is a beta-catenin destruction complex scaffold protein and a transcription factor of the Wnt signaling pathway. Variation in the Wnt signaling pathway has been associated with the formation of some primary human cancers. Axin-1 mutation has been linked to oral squamous cell carcinoma. Axin-1 binds to GSK3alpha and activates the Wnt signaling pathway which is shown to be associated with growth and development of cells. Axin-1 has also been implicated in Alzheimer′s disease.

Immunogen

KLH-conjugated linear peptide corresponding to human Axin-1.

Application

Anti-Axin-1 Antibody is an antibody against Axin-1 for use in WB, IC, IH.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Axin-1 in A431 and HeLa cells.

Immunohistochemistry Analysis: 1:1,000 dilution from a representative lot detected Axin-1 in human kidney and human skin tissues.

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Axin-1 on 10 µg of HEK293 cell lysate.

Target description

~110 kDa observed. An uncharacterized band appears at ~40 and 45 kDa in some lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiwei Zhou et al.
Pharmaceutics, 13(3) (2021-04-04)
Genetic evidence has indicated that β-catenin plays a vital role in glucose and lipid metabolism. Here, we investigated whether pyrvinium, an anthelmintic agent previously reported as a down-regulator of cellular β-catenin levels, conferred any metabolic advantages in treatment of metabolic
Yiwei Li et al.
Cell stem cell, 28(1), 63-78 (2020-10-15)
Enormous amounts of essential intracellular events are crowdedly packed inside picoliter-sized cellular space. However, the significance of the physical properties of cells remains underappreciated because of a lack of evidence of how they affect cellular functionalities. Here, we show that
Anuradha Kumari et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(4), e21539-e21539 (2021-03-21)
An aberrant accumulation of nuclear β-catenin is closely associated with the augmentation of cancer malignancy. In this work, we report that several microtubule-targeting agents (MTAs) such as vinblastine, taxol, and C12 (combretastatin-2-aminoimidazole analog) inhibit Wnt/β-catenin signaling in oral squamous cell
Ludovic Zimmerlin et al.
Development (Cambridge, England), 143(23), 4368-4380 (2016-11-02)
The derivation and maintenance of human pluripotent stem cells (hPSCs) in stable naïve pluripotent states has a wide impact in human developmental biology. However, hPSCs are unstable in classical naïve mouse embryonic stem cell (ESC) WNT and MEK/ERK signal inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service