Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA022914

Sigma-Aldrich

Anti-DVL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DSH homolog 2, Anti-Dishevelled-2, Anti-Segment polarity protein dishevelled homolog DVL-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

FNGRVVSWLVSSDNPQPEMAPPVHEPRAELAPPAPPLPPLPPERTSGIGDSRPPSFHPNVSSSHENLEPETETESVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DVL2(1856)

Immunogen

Segment polarity protein dishevelled homolog DVL-2 recombinant protein epitope signature tag (PrEST)

Application

Anti-DVL2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

DVLs (dishevelled segment polarity proteins) are cytoplasmic proteins, which are needed for embryonic development, cell differentiation and tumor formation. They are key players in canonical (β-catenin-dependent) as well as non-canonical Wnt pathways. DVL2 is associated with prostate cancer. The gene DVL2 is mapped to human chromosome 17p13.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75518

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinhui Yang et al.
Molecular biology reports, 40(7), 4241-4250 (2013-05-09)
To identify Dishevelled-2 (Dvl2) is a prostate cancer-associated gene and analyze the effects on the growth and invasive capacity of human prostate cancer (PCa) cells. Dvl2 mRNA expression was measured in PCa cell lines and tissue samples, by real-time reverse
Dong Chen et al.
Molecular medicine reports, 11(3), 2092-2096 (2014-11-15)
Hirschsprung's disease (HSCR) is a congenital disorder characterized by an absence of enteric ganglion cells in the terminal regions of the gut during development. To date, the cause of HSCR remains unclear, although the pathogenesis of this complex disease is
Fong Chun Chan et al.
Blood, 125(6), 959-966 (2014-11-15)
Effective treatment of diffuse large B-cell lymphoma (DLBCL) is plagued by heterogeneous responses to standard therapy, and molecular mechanisms underlying unfavorable outcomes in lymphoma patients remain elusive. Here, we profiled 148 genomes with 91 matching transcriptomes in a DLBCL cohort
Ikuko Takeda et al.
Pathobiology : journal of immunopathology, molecular and cellular biology, 81(2), 94-99 (2014-01-25)
Gene expression patterns differ in the two types of skeletal muscle fiber. The Wnt signaling pathway, which includes low-density lipoprotein receptor-related protein 6 (LRP6), has been associated with cell differentiation and glucose metabolism in skeletal muscles. We examined the relationships
Magdalena Banach-Orlowska et al.
FEBS letters, 589(4), 532-539 (2015-01-28)
APPL1 is a multifunctional endocytic adaptor which acts at different steps of various signaling pathways. Here we report that APPL1 interacts with Dvl2, a protein known to activate the canonical and non-canonical Wnt pathways. APPL1 synergizes with Dvl2 and potentiates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service