Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1945

Sigma-Aldrich

Anti-Vitronectin Antibody, clone BV2

clone BV2, Chemicon®, from mouse

Synonym(s):

Serum spreading factor, S-protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BV2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VTN(7448)

Related Categories

Specificity

Human vitronectin. Specificity determined by ELISA and immunoprecipitation of I125 labeled vitronectin purified from plasma (Zanetti et al., 1994). The antibody reacts with vitronectin both in its native and denatured form.

Immunogen

Purified human plasma vitronectin (Preissner, 1985).

Application

Anti-Vitronectin Antibody, clone BV2 is an antibody against Vitronectin for use in ELISA, FC, IP, WB, IH.
ELISA

Immunoprecipitation

Western blot

Immunohistochemistry/immunofluorescence

Inhibits endothelial cell adhesion and spreading on vitronectin-coated surfaces.

Flow Cytometry

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Purified immunoglobulin by Protein A chromatography. Liquid in PBS containing 0.05% sodium azide and 0.1% BSA.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Cantini et al.
Advanced biosystems, 1(9), 1700047-1700047 (2018-03-03)
Surface functionalization strategies of synthetic materials for regenerative medicine applications comprise the development of microenvironments that recapitulate the physical and biochemical cues of physiological extracellular matrices. In this context, material-driven fibronectin (FN) nanonetworks obtained from the adsorption of the protein
Aceruloplasminemia: retinal histopathologic manifestations and iron-mediated melanosome degradation.
Wolkow, N; Song, Y; Wu, TD; Qian, J; Guerquin-Kern, JL; Dunaief, JL
Archives of Ophthalmology (1960)
Asa Thulin et al.
Molecular cancer research : MCR, 7(11), 1792-1802 (2009-11-12)
The angiogenesis inhibitor histidine-rich glycoprotein (HRG) constitutes one of several examples of molecules regulating both angiogenesis and hemostasis. The antiangiogenic properties of HRG are mediated via its proteolytically released histidine- and proline-rich (His/Pro-rich) domain. Using a combination of immunohistochemistry and
Vitronectin in human hepatic tumours contributes to the recruitment of lymphocytes in an alpha v beta3-independent manner.
Edwards, S; Lalor, PF; Tuncer, C; Adams, DH
British Journal of Cancer null
Marie Monet-Leprêtre et al.
Brain : a journal of neurology, 136(Pt 6), 1830-1845 (2013-05-08)
Cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy, or CADASIL, one of the most common inherited small vessel diseases of the brain, is characterized by a progressive loss of vascular smooth muscle cells and extracellular matrix accumulation. The disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service