Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04-773

Sigma-Aldrich

Anti-phospho-MYPT1 (Thr850) Antibody,, rabbit monoclonal

culture supernatant, clone SA19, from rabbit

Synonym(s):

Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, Protein phosphatase myosin-binding subunit, myosin phosphatase, target subunit 1, protein phosphatase 1, regulatory (inhibitor) subunit 12A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SA19, monoclonal

species reactivity

rat, chicken, mouse, human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr850)

Gene Information

human ... PPP1R12A(4659)

General description

Phosphorylation of myosin is crucial to smooth muscle contraction. The degree of myosin phosphorylation is dependent in part by the activity of myosin phosphatase. Recent findings suggest that this enzyme can be regulated, which has lead to interest in characterizing this phosphatase. Myosin phosphatase seems to be comprised of three subunits (catalytic subunit of type 1 phosphatase-δ (PP1) and two non-catalytic subunits, large and small (M20)). Studies suggest that the large subunit is a targeting subunit and is termed myosin phosphatase target subunit (MYPT). MYPT has been shown to be a substrate for the Rho-associated kinase (ROK). ROK mediates an agonist-induced increase in myosin phosphorylation and force by inhibiting myosin light chain phosphatase activity through phosphorylation of MYPT.

Specificity

Predicted to cross-react with mouse and rat.
Recognizes MYPT1 phosphorylated on Thr850, doublet at Mr ~130 kDa. A non-specific protein was also detected, Mr ~55-60 kDa.

Immunogen

Epitope: pThr850(Ch) or pThr853(H)
KLH-conjugated, synthetic peptide containing the sequence …RS[pT]GV…, in which [pT] corresponds to phosphorylated threonine at amino acid residue 850 of chicken myosin phosphatase target subunit 1 (MYPT1). Corresponds to Thr853 in human MYPT1.

Application

Anti-phospho-MYPT1 (Thr850) Antibody, detects level of phospho-MYPT1 (Thr850) & has been published & validated for use in WB.

Quality

Westerin Blot Analysis: A 1:500-1:1000 dilution of this antibody detected MYPT1 in RIPA lysates from HEK293 cells phosphorylated by preincubation with ROKα/ROCK-II, active (Catalog # 14-338).

Target description

Doublet at ~130 kDa

Linkage

Replaces: 05-773

Analysis Note

Control
HEK293 cell lysate phosphorylated by ROKα/ROCKII.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ebbe Boedtkjer et al.
Circulation, 124(17), 1819-1829 (2011-09-29)
Disturbances in pH affect artery function, but the mechanistic background remains controversial. We investigated whether Na(+), HCO₃- transporter NBCn1, by regulating intracellular pH(pH₁), influences artery function and blood pressure regulation. Knockout of NBCn1 in mice eliminated Na+, HCO₃⁻ cotransport and
E V Bouzinova et al.
Acta physiologica (Oxford, England), 224(1), e13059-e13059 (2018-02-27)
The Na,K-ATPase is involved in a large number of regulatory activities including cSrc-dependent signalling. Upon inhibition of the Na,K-ATPase with ouabain, cSrc activation is shown to occur in many cell types. This study tests the hypothesis that acute potentiation of
Vincent Sauzeau et al.
Methods in molecular biology (Clifton, N.J.), 1527, 213-218 (2017-01-25)
Increased arterial tone and the resulting rise in peripheral vascular resistance are major determinants of the elevated arterial pressure in hypertension. The RhoA/Rho kinase signaling pathways are now recognized as a major regulator of vascular smooth muscle contraction and arterial
Huaqing Zheng et al.
Acta pharmaceutica Sinica. B, 11(12), 3994-4007 (2022-01-14)
Vascular smooth muscle cell (VSMC) migration plays a critical role in the pathogenesis of many cardiovascular diseases. We recently showed that TMEM16A is involved in hypertension-induced cerebrovascular remodeling. However, it is unclear whether this effect is related to the regulation
Aditi U Gurkar et al.
Nature communications, 4, 2189-2189 (2013-07-24)
The Ser/Thr Rho kinase 1 (ROCK1) is known to have major roles in a wide range of cellular activities, including those involved in tumour metastasis and apoptosis. Here we identify an indispensable function of ROCK1 in metabolic stress-induced autophagy. Applying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service