Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V-029

Supelco

Valproic Acid-D6 solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Valproic Acid-D6 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10D6O2
CAS Number:
Molecular Weight:
150.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC(C(CCC([2H])([2H])[2H])CCC([2H])([2H])[2H])=O

InChI

1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/i1D3,2D3

InChI key

NIJJYAXOARWZEE-WFGJKAKNSA-N

General description

An internal standard and Certified Spiking Solution® suitable for quantitation of valproic acid levels in urine or other matrices by LC/MS or GC/MS for testing applications in clinical toxicology, forensic analysis, medication monitoring, urine drug testing, or therapeutic drug monitoring. Valproic acid, sold under trade names such as Depakote®, Valparin, or Stavzor®, is an antiepileptic drug and mood stabilizer. The drug is prescribed for treatment of epilepsy, bipolar disorder, migraine headaches, and depression.

Application


  • Catalysis driven by an amyloid-substrate complex: Research demonstrates the role of buffer concentrates in maintaining optimal pH and stability conditions during biochemical reactions involving amyloid substrates, critical for developing new therapeutic agents (Sawazaki et al., 2024).

  • High throughput microfluidics-based synthesis of PEGylated liposomes for precise size control and efficient drug encapsulation: This research utilizes buffer concentrates to manage the pH and ionic strength during the microfluidic synthesis of PEGylated liposomes, critical for achieving size precision and high encapsulation efficiency in drug delivery systems (Akar et al., 2024).

  • Development of Analytical Quality by Design RP-HPLC Method and Its Validation for Estimation of Gefitinib from Bulk, Tablet Dosage Form and Complex Nanoformulation: Buffer concentrates play a vital role in this study, optimizing the RP-HPLC method for precise quantification of gefitinib, a critical aspect in ensuring the quality and efficacy of pharmaceutical products (More et al., 2024).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Depakote is a registered trademark of Sanofi S.A.
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Stavzor is a registered trademark of Noven Therapeutics, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service