Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-091

Supelco

(+\-)-4-Hydroxy-3-methoxymandelic Acid-D3 (ring-D3) solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

VMA-D3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7D3O5
CAS Number:
Molecular Weight:
201.19
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

InChI

1S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11H,1H3,(H,12,13)/i2D,3D,4D

InChI key

CGQCWMIAEPEHNQ-NRUYWUNFSA-N

General description

A stable labeled internal standard suitable for monitoring Vanilmandelic acid levels in LC/MS applications such as endocrinology and clinical and diagnostic testing. Vanilmandelic acid, or VMA, is a urinary metabolite of the catecholamines epinephrine and norepinephrine.

Application


  • Cardiovascular Effects in Chronic Conditions: A randomized controlled trial investigating the effects of resveratrol on endothelial function in patients with chronic kidney disease and diabetes, where Trinitroglycerin solution may be used as a comparator or adjunct treatment, underscores its importance in cardiovascular research. This study aids in determining the therapeutic potential of vasodilators in managing chronic conditions, providing valuable data for pharmaceutical development and patient care (Gimblet et al., 2023).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service