Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T-186

Supelco

Taxifolin

new

certified reference material, vial of 1 mL, Cerilliant®

Synonym(s):

Taxifolin, (2R,3R)-3,3′,4′,5,7-Pentahydroxyflavanone, (2R,3R)-Dihydroquercetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O7
CAS Number:
Molecular Weight:
304.25
Beilstein:
5299277
MDL number:

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 1 mL

manufacturer/tradename

Cerilliant®

format

neat

storage temp.

-10 to -25°C

SMILES string

O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3

InChI

1S/C15H12O7/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6/h1-5,14-19,21H/t14-,15+/m0/s1

InChI key

CXQWRCVTCMQVQX-LSDHHAIUSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service