Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-144

Supelco

Silybum Mix solution

200 μg/mL in methanol ((each analyte)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22O10
Molecular Weight:
482.44
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Isosilybin, 200 μg/mL
Silybin, 200 μg/mL
Silychristin, 200 μg/mL
Silydianin, 200 μg/mL
Taxifolin
Taxifolin, 200 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

200 μg/mL in methanol ((each analyte))

format

multi-component solution

storage temp.

-10 to -25°C

General description

This phytochemical certified reference materials mix is produced at the Millipore Sigma Cerilliant site in Round Rock Texas under ISO/IEC 17025 and ISO 17034 accreditation. A comprehensive certificate including traceable values and expanded uncertainties for all the analytes is available.


Discover the entire range of phytochemical certified reference material solutions from Cerilliant

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service