Skip to Content
MilliporeSigma
All Photos(1)

Documents

P-179

Supelco

Phosphatidylethanol 16:0/18:1 (PEth 16:0/18:1) solution

1 mg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H74O8P · NH4
CAS Number:
Molecular Weight:
720.01
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in methanol ((as free base))

format

single component solution

storage temp.

-65 to -96°C

General description

PEth 16:0/18:1, also known as phosphatidylethanol 16:0/18:1, is a sensitive and specific biomarker for alcohol use and abuse. Since the detection window of PEth 16:0/18:1 in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/18:1 levels is routinely performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing. PEth 16:0/18:1 has also been investigated as a biomarker to confirm prenatal alcohol exposure.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service