Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-082

Supelco

Propofol β-D-glucuronide sodium salt

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Propofol beta-D-glucuronide sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NaO7
Molecular Weight:
376.38
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=C2C(NC(CCOCCC(O)=O)=O)=CC=C1)N([C@H]3CCC(NC3=O)=O)C2=O.O=C(CCOCCC(O)=O)N[C@@H](C(C)(C)C)C(N4C[C@H](O)C[C@H]4C(NCC5=CC=C(C6=C(C)N=CS6)C=C5)=O)=O

InChI

1S/C18H26O7.Na/c1-8(2)10-6-5-7-11(9(3)4)15(10)24-18-14(21)12(19)13(20)16(25-18)17(22)23;/h5-9,12-14,16,18-21H,1-4H3,(H,22,23);/q;+1/p-1/t12-,13-,14+,16-,18+;/m0./s1

InChI key

RPECYLUOLHOVLR-VVDXHYSNSA-M

General description

Propofol glucuronide is the primary urinary metabolite of propofol, marketed as the anesthetic Diprivan® for sedation and the induction and maintenance of general anesthesia. According to DEA′s Drugs and Chemicals of Concern list, propofol abuse typically involves recreational use by anesthetists, practitioners, and other health care staff.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Diprivan is a registered trademark of Abraxis Bioscience, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service