Skip to Content
MilliporeSigma
All Photos(1)

Documents

O-008

Supelco

Oxycodone-D6 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15NO4D6
CAS Number:
Molecular Weight:
321.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O)C([2H])([2H])[2H]

InChI

1S/C18H21NO4/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10/h3-4,13,16,21H,5-9H2,1-2H3/t13-,16+,17+,18-/m1/s1/i1D3,2D3

InChI key

BRUQQQPBMZOVGD-UKHJWGAOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard suited for quantitation of oxycodone levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Oxycodone is a semi-synthetic opioid analgesic sold under many trade names including OxyContin® and Percocet® for treatment of moderate to severe pain. Oxycodone is also sold illicitly under street names such as "Roxies," "Blues," and "Perks".

Application


  • Oxycodone-D₆ as an isotopic standard: Oxycodone-D₆ is employed extensively as a high-quality isotopic standard in analytical chemistry, particularly in mass spectrometry. This application is crucial for ensuring accuracy and precision in the quantification of oxycoDonein various biological matrices, enhancing drug monitoring and forensic analyses.

  • Stable isotope-labeled oxycoDonefor pharmacokinetic studies: Researchers use Oxycodone-D₆ to investigate the pharmacokinetics of oxycodone, including its absorption, distribution, metabolism, and excretion. The use of this stable isotope-labeled compound allows for more precise tracking of drug metabolites, contributing significantly to the understanding of drug behavior in the human body.

  • Oxycodone-D₆ in metabolite analysis: The deuterated form of oxycoDoneis critical in metabolite profiling studies. It serves as an internal standard that facilitates the identification and quantification of oxycoDonemetabolites, thereby aiding in the exploration of metabolic pathways and potential drug interactions.

  • Use in analytical standard development for biochemistry research: Oxycodone-D₆ solution is integral to developing robust analytical methods for drug testing and research. Its stability and reliability make it a preferred choice for validating new analytical techniques in biochemistry labs.

  • Investigation of opioid receptor binding assays: This compound is utilized in studies exploring opioid receptor interactions, aiding in the development of targeted therapies for pain management. Oxycodone-D₆ helps in understanding the binding efficiency and pharmacodynamics of opioids at the molecular level.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
OxyContin is a registered trademark of Purdue Pharma L.P.
Percocet is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service