Skip to Content
MilliporeSigma
All Photos(1)

Documents

A-128

Supelco

Amitriptyline N-ß-D-glucuronide solution

100 μg/mL in acetonitrile: water (1:1), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31NO6
CAS Number:
Molecular Weight:
453.53
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile: water (1:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

InChI

1S/C26H31NO6/c1-27(2,25-23(30)21(28)22(29)24(33-25)26(31)32)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20/h3-6,8-12,21-25,28-30H,7,13-15H2,1-2H3/t21-,22-,23+,24-,25+/m0/s1

InChI key

WXMXRAPAGYPAJI-ARXROMJUSA-N

General description

A certified solution standard of Amitriptyline N-ß-D-glucuronide, a metabolite of the tricyclic antidepressant amitriptyline. Sold under trade names such as Amitrip, Elavil, and Levate, amitriptyline is prescribed to treat a number of mental disorders ranging from major depressive disorder and anxiety disorder to bipolar disorder. This solution-based Certified Reference Material is applicable for use in LC/MS or GC/MS testing applications in clinical toxicology, forensic analysis, workplace drug testing, and pharmaceutical research.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service