G-007
Gabapentin solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
About This Item
Recommended Products
grade
certified reference material
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
1.0 mg/mL in methanol
application(s)
pharmaceutical (small molecule)
format
single component solution
storage temp.
−20°C
SMILES string
NCC1(CCCCC1)CC(O)=O
InChI
1S/C9H17NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-7,10H2,(H,11,12)
InChI key
UGJMXCAKCUNAIE-UHFFFAOYSA-N
Gene Information
human ... CACNA1A(773) , CACNA1B(774) , CACNA1C(775) , CACNA1D(776) , CACNA1E(777) , CACNA1F(778) , CACNA1G(8913) , CACNA1H(8912) , CACNA1I(8911) , CACNA1S(779) , CACNA2D1(781) , CACNA2D2(9254) , CACNA2D3(55799) , CACNA2D4(93589) , CACNB1(782) , CACNB2(783) , CACNB3(784) , CACNB4(785) , CACNG1(786) , CACNG2(10369) , CACNG3(10368) , CACNG4(27092) , CACNG5(27091) , CACNG6(59285) , CACNG7(59284) , CACNG8(59283)
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service