Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P-008

Supelco

Phenobarbital solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

5-Ethyl-5-phenylbarbituric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

CCC1(C(=O)NC(=O)NC1=O)c2ccccc2

InChI

1S/C12H12N2O3/c1-2-12(8-6-4-3-5-7-8)9(15)13-11(17)14-10(12)16/h3-7H,2H2,1H3,(H2,13,14,15,16,17)

InChI key

DDBREPKUVSBGFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenobarbital is the oldest and most widely used barbiturate. The drug is an anticonvulsant approved for treatment of all seizure types with the exception of absence seizures. This certified solution standard is suitable as starting material for the preparation of calibrators and controls in phenobarbital testing by GC/MS or LC-MS/MS.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuzhen Liu et al.
Breast cancer research and treatment, 149(2), 439-448 (2015-01-02)
PAM50-defined breast cancer intrinsic subtypes and risk-of-relapse (ROR) scores are prognostic and predictive of endocrine therapy and some chemotherapy. We investigated the prognostic and predictive effect of PAM50 classifications by chemotherapy type. NCIC CTG MA.21 randomized 2,104 patients to doxorubicin
Patrick Kwan et al.
Epilepsia, 45(9), 1141-1149 (2004-08-27)
Phenobarbital (PB) is the most widely used antiepileptic drug (AED) in the developing world and remains a popular choice in many industrialized countries. Meta-analyses of randomized controlled trials suggest that few differences in efficacy exist between PB and other established
M Lundqvist et al.
Acta paediatrica (Oslo, Norway : 1992), 102(9), 863-867 (2013-06-07)
Treatment of neonatal seizures still relies primarily on phenobarbital, despite an estimated efficacy of less than 50% and concern over neurodegenerative side effects. The objective of this study was to evaluate the efficacy and safety of lidocaine as second-line treatment
Susumu Kodama et al.
Drug metabolism reviews, 38(1-2), 75-87 (2006-05-11)
In the early 1960s, phenobarbital (PB) was shown to induce hepatic drug metabolism and the induction was implicated in the molecular mechanism of drug tolerance development. Since then, it has become evident that PB not only induces drug metabolism, but
L Corcos et al.
Pharmacology & toxicology, 89(3), 113-122 (2001-10-09)
Phenobarbital has long been used as a sedative and antiepileptic drug. The drug is the representative of a myriad of lipophilic molecules able to evoke a pleiotropic response in the liver and also in prokaryotes and flies. A great deal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service