Skip to Content
MilliporeSigma
All Photos(1)

Documents

D-068

Supelco

Duloxetine-D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D3H16NOS · HCl
Molecular Weight:
336.89
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.[2H]C([2H])([2H])NCC[C@H](Oc1cccc2ccccc12)c3cccs3

InChI

1S/C18H19NOS.ClH/c1-19-12-11-17(18-10-5-13-21-18)20-16-9-4-7-14-6-2-3-8-15(14)16;/h2-10,13,17,19H,11-12H2,1H3;1H/t17-;/m0./s1/i1D3;

InChI key

BFFSMCNJSOPUAY-HQKLNPQUSA-N

General description

A stable-labeled internal standard suitable for numerous GC/MS and LC/MS applications from clinical toxicology and urine drug testing to forensic analysis and isotope dilution methods. Duloxetine, marketed in the US under the trade name Cymbalta®, is a serotonin-norepinephrine reuptake inhibitor (SNRI) used to treat major depressive disorder and to alleviate pain associated with diabetic neuropathy, fibromyalgia, and osteoarthritis.

Application


  • Rapid Structure-Based Screening for COVID-19: Duloxetine-D₃ hydrochloride solution plays a key role in rapid structure-based screening, potentially informing the development of therapeutic agents for COVID-19 due to its properties as a serotonin-norepinephrine reuptake inhibitor, crucial for high-impact pharmaceutical research (Yang et al., 2020).

  • UV-Triggered Drug Release: The high-purity Duloxetine-D₃ solution enhances UV-triggered drug release studies from mesoporous titanium nanoparticles loaded with pharmaceutical agents, indicating significant benefits for developing controlled drug delivery systems in biotechnological applications (Zuo et al., 2024).

  • Antioxidant Activity and Eczema Treatment: Duloxetine-D₃ HCl serves as an internal standard in studies evaluating the antioxidant activities and therapeutic applications of drug-loaded liposomes for eczema treatment, highlighting its role in enhancing bioanalytical assay development (Shen et al., 2024).

  • Synthesis and Application of EGFR Inhibitors for Cancer: As a stable isotope-labeled compound, Duloxetine-D₃ hydrochloride is used in the synthesis and clinical application studies of small-molecule EGFR inhibitors, underlining its critical application in cancer therapy research (Wang et al., 2024).

  • Nanoparticle Drug Release Behaviors: The solution′s role in studying poly(lactic-co-glycolic acid) nanoparticles highlights its utility in pharmaceutical-grade calibration for analyzing hydrophilic and hydrophobic drug release behaviors, essential for pharmaceutical and biomedical research (Wang et al., 2024).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Cymbalta is a registered trademark of Eli Lilly and Co.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service