Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-091

Supelco

Clozapine-D4 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D4H15ClN4
CAS Number:
Molecular Weight:
330.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN(CC1([2H])[2H])CC([2H])([2H])N1C2=NC(C=C(Cl)C=C3)=C3NC4=CC=CC=C24

InChI

1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3/i10D2,11D2

InChI key

QZUDBNBUXVUHMW-MKQHWYKPSA-N

General description

A stable-labled internal standard for quantitation of Clozapine levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, forensic analysis, and urine drug testing applications. Clozapine, sold as Clozaril® and FazaClo® in the US, is an atypical antipsychotic drug used for treatment of schizophrenia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Clozaril is a registered trademark of Sandoz, Inc.
FazaClo is a registered trademark of Jazz Pharmaceuticals International III Limited
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service