Skip to Content
MilliporeSigma

A-085

Amitriptyline-d3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Amitriptyline-d3 hydrochloride solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20D3H21ClN
CAS Number:
Molecular Weight:
316.88
MDL number:
UNSPSC Code:
41116107
EC Number:
200-659-6
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.N(CCC=C1c2c(cccc2)CCc3c1cccc3)(C([2H])([2H])[2H])C

InChI

1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H/i1D3;

InChI key

KFYRPLNVJVHZGT-NIIDSAIPSA-N

General description

A new internal standard suitable for a wide variety of uses in LCMS or GCMS applications from forensic or clinical toxicology analysis to urine drug testing and isotope dilution methods. Amitriptyline, marketed as the brand drug Elavil, Tryptizol, or Laroxyl, is a tricyclic antidepressant used in the treatment of major depression, clinical/endogenous depression and involutional melancholia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class

3 - Flammable liquids

WGK

WGK 1

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library