Skip to Content
MilliporeSigma
All Photos(1)

Documents

880229P

Avanti

DSPE-PEG(2000)-DBCO

Avanti Research - A Croda Brand

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[dibenzocyclooctyl(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (880229P-10mg)
pkg of 1 × 100 mg (880229P-100mg)
pkg of 1 × 25 mg (880229P-25mg)
pkg of 1 × 50 mg (880229P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

polymerizable lipids
click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\NC(CCC(N1CC(C=CC=C2)=C2C#CC3=C1C=CC=C3)=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C

General description

DSPE-PEG(2000)-DBCO is a functionalized polyethylene glycol (PEG) lipid conjugate.

Application

DSPE-PEG(2000)-DBCO has been used in the preparation of peptide conjugated fluorescent nanoparticle by modified thin-film hydration technique. It has also been used for homeotropic anchoring of the mesogen molecules.

Packaging

5 mL Amber Glass Screw Cap Vial (880229P-100mg)
5 mL Amber Glass Screw Cap Vial (880229P-10mg)
5 mL Amber Glass Screw Cap Vial (880229P-25mg)
5 mL Amber Glass Screw Cap Vial (880229P-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Active nematic emulsions
Guillamat BP, et al.
Science Advances, 4(4) (2018)
Active nematic emulsions
Guillamat BP, et al.
Science advances, 4(4) (2018)
A novel nanoparticle delivery system for targeted therapy of noise-induced hearing loss
Kayyali MN, et al.
J. Controlled Release, 279, 243-250 (2018)
Patty Wong et al.
Bioconjugate chemistry (2020-01-22)
Lipid nanodiscs (LNDs), comprising a phospholipid bilayer encircled by two molecules of a recombinant membrane scaffold protein, can be targeted to tumors with covalently attached antibodies (Abs) or their fragments. Antibody attachment to click chemistry based PEGylated lipids on LNDs
John C Jewett et al.
Journal of the American Chemical Society, 132(11), 3688-3690 (2010-03-02)
Bioorthogonal chemical reactions, those that do not interact or interfere with biology, have allowed for exploration of numerous biological processes that were previously difficult to study. The reaction of azides with strained alkynes, such as cyclooctynes, readily forms a triazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service