Z742868
Kern PLS series precision balances
Kern PLS 720-3A, weighing capacity 720 g, accuracy: 0.001 g, precision: ±0.002 g
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
All Photos(1)
About This Item
UNSPSC Code:
41111502
NACRES:
NB.71
Recommended Products
AC/DC input
110 - 240 V AC, universal plug set
packaging
pack of 1 ea
manufacturer/tradename
Kern PLS 720-3A
weighing capacity measuring range
720 g, accuracy: 0.001 g, precision: ±0.002 g
Related Categories
General description
Range of precision balances with CAL adjusting program for quick setting of the balance accuracy.
Ergonomically optimised keypad for left and righthanded users
Ergonomically optimised keypad for left and righthanded users