Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z742495

MyBlock HL Mini dry bath with heated lid

AC/DC input 100 V / 240 V AC, EuroPlug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NC.21

feature

accuracy± 0.3 °C
incremental 0.1 °C
temperature uniformity± 0.2 °C

packaging

pkg of 1

manufacturer/tradename

Benchmark Scientific BSH200-HL-E

parameter

+5 °C temp. range (100 °C)

W × D × H

4 in. (10.5 cm) × 6 in. (15 cm) × 4.5 in. (11.5 cm)

block size

1.85 in. (4.7 cm) × 2.8 in. (7.1 cm) × 1.25 in. (3.2 cm)

capacity

, holds 1 block

weight

1 kg (2.2 lb)

Looking for similar products? Visit Product Comparison Guide

General description

The new MyBlock HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation.
Both the block and lid temperature can be selected independently in increments of 0.1°C, making this MyBlock the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions & other temperature hyper-sensitive procedures.

  • Heated lid prevents evaporation & condensation
  • Compact, fits in the palm of your hand
  • Independant temperature control of lid & block
  • Exchangeable blocks for 0.2 to 0.5ml tubes
  • Room temperature to 37°C in just 2 minutes

Legal Information

MyBlock is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service