Key Documents
Z740712
Sigma-Aldrich
Aldrich® Essentials boiling flask, round-bottom, ST/NS 24/29
capacity 50 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass flask
round bottom
single-neck
packaging
pkg of 1 ea
capacity
50 mL
General description
Clear borosilicate glass, round bottomed boiling flasks with outer ST ground joint neck. White marking spot.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)