Key Documents
Z515647
Sigma-Aldrich
Aldrich® evaporating flask, pear-shaped
capacity 1,000 mL, joint: ST/NS 29/32, non-coated, heavy wall
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
VB.02
material
borosilicate glass
long
pear-shaped
single-neck
Joint
joint: ST/NS29/32
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Büchi equivalent, 00435
capacity
1,000 mL
General description
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)