Key Documents
Z723061
Sigma-Aldrich
Aldrich® Erlenmeyer flask with ST joint
capacity 125 mL, joint: ST/NS 24/40
Synonym(s):
conical flasks, glass flasks, lab flask, laboratory flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom
Joint
joint neck24/40
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
without cap
packaging
case of 1
manufacturer/tradename
Sigma-Aldrich
capacity
125 mL
Other Notes
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)