Key Documents
SYNF303450
Synthware single-neck round-bottom flask
500 mL, joint: ST/NS 34/45
Synonym(s):
Synthware F303450, glass round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS34/45
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F303450
capacity
500 mL
500 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)