Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z680028

MyBlock mini dry bath block

to hold, 40 X 0.2 Ml Tubes Or 5 X x dry strips

Synonym(s):

block dry, dry baths, dry blocks, heat blocks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NB.76

feature

to hold 40 X 0.2 Ml Tubes Or 5 X x dry strips

manufacturer/tradename

Benchmark Scientific BSH100-02

technique(s)

PCR: suitable

Looking for similar products? Visit Product Comparison Guide

General description

To fit MyBlock mini dry bath/block heater.

Legal Information

MyBlock is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service