MilliporeSigma
All Photos(4)
Key Documents
Z635383
Swiss Boy lab jacks
load ≤90 kg (middle position), platform L × W 240 mm × 320 mm, Model 118
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
manufacturer/tradename
Grauer
parameter
≤90 kg load (middle position)
120-500 mm vertical limit
platform L × W
240 mm × 320 mm
color
gray
General description
Jack can be raised with load in place.
These high quality Swiss made jacks have massive cast aluminum frames with a chemically resistant anodised finish. Swiss boy jacks are designed to carry heavy loads safely. Available either in standard size or with extended carrying plates for large pots or pans. Units carrying more that 90kg are also available with a motorized lift adjustment for enhanced convenience. The max load is for the middle position. Supplied with top and bottom plates.
Products with motorized jacks are available in Europe only.