Key Documents
Z158062
Sigma-Aldrich
Poly-jaque lab jack
polypropylene, plate size 6 in. × 6 in.
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
polypropylene
manufacturer/tradename
Bel-Art
plate size
6 in. × 6 in.
General description
Nonsparking and nonmagnetic, corrosion-resistant. Self-lubricating with polyether-imide/PTFE bushings. Working height to 11in. Maximum static load is 15lb.
Legal Information
Poly-jaque is a trademark of SP Scienceware, a division of SP