Key Documents
Z543586
Sigma-Aldrich
Delivery cap for VerSA-Flow dispenser
three-hole
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
PBT
PFA
PTFE
O.D.
1/8 in.
3.2 mm
fitting
GL 45 female
General description
Caps require a GL-45 bottle adapter such as Z332305 for use with Aldrich 2 L and 4 L bottles.
Legal Information
VerSA-Flow is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Related Content
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.