Documents
SYNA352440
Synthware distillation adapter
top joint: ST/NS 24/40, bottom joint: ST/NS 24/40, side joint: 24/40, side arm angle: 75 deg
Synonym(s):
Synthware A352440, adapter, sidearm adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
bottom joint: ST/NS24/40
side joint: 24/40
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A352440
General description
For use as a component in atmospheric or vacuum distillations.
Legal Information
Synthware is a trademark of Kemtech America Inc.