Key Documents
Z515523
Sigma-Aldrich
Aldrich® evaporating flask, pear-shaped
capacity 25 mL, joint: ST/NS 14/20, non-coated, heavy wall
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
long
pear-shaped
single-neck
Joint
joint: ST/NS14/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
capacity
25 mL
General description
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC