Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z509019

Sigma-Aldrich

AldraSORB water trapping packets

medium, volume 200-1,000 mL

Synonym(s):

AldraSORB Packets, Trapping Packets, Water Trapping Packets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24141516
NACRES:
NB.43

volume

200-1,000 mL

weight

10 g

Looking for similar products? Visit Product Comparison Guide

General description

The AldraSorb packets are made from an ultra-high molecular weight polyethylene, electronic grade, hydrophobic membrane that contains pharmaceutical quality molecular sieves. The full filtration capability of the molecular particulates is maintained and the surrounding solvent is not compromised. Packets are vacuum sealed.

  • Maintains low levels of water impurities for up to 3 months
  • Ideal for solid-phase and solution-phase peptide synthesis
  • Effectively dries solvents to below 50 ppm

Note: These traps are not recommended for solvents having more than 0.05% or 500 ppm of water. They should not be used in acidic solvents.

Legal Information

AldraSORB is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service