Key Documents
Z287253
Sigma-Aldrich
Aldrich® low-pressure basic three station manifold
hose outlet I.D. 1/4 in.
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
hose outlet I.D.
1/4 in.
General description
This system offers complete manifold installation flexibility.
Made of brass with 1/4 in. NPTM inlet. Consists of three stations, each with a needle valve and 1/4 in. I.D. hose barb outlet.
As increased manifold capacity is needed, simply order the required parts and install them on-site (refer to ′Related Products′ tab).
Caution
Do not exceed 200 psi inlet gas pressure.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC