Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z273627

Solid-glass beads

borosilicate, diam. 2 mm

Synonym(s):

borosilicate beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102422
NACRES:
NB.43
Pricing and availability is not currently available.

material

borosilicate

packaging

pack of 1 ea

diam.

2 mm

Looking for similar products? Visit Product Comparison Guide

General description

For use as distillation column packing, mixing beads, and boiling stones. Each 454 g box has a packing volume of approx. 360 cm3. +/-10% variation in bead size.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service