Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z272779

Sigma-Aldrich

Nalgene® PFA beaker

capacity 250 mL

Synonym(s):

chemical resistance beaker, plastic beaker

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43
Pricing and availability is not currently available.

material

PFA

packaging

pack of 1 ea

manufacturer/tradename

Nalgene 1510-0250

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

General description

PFA unaffected by most chemicals. Withstands a temperature range from -270 to 250°C.

Other Notes

Browse all plastic beakers

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service