Documents
Z154407
Ace Trubore® PTFE 4-piece stirrer bearing
joint: ST/NS 24/40, fits shaft o.d., 10 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
Joint
joint: ST/NS24/40
feature
fits shaft o.d. 10 mm
manufacturer/tradename
Ace Glass 806643
General description
Self-aligning, lubricant-free PTFE Trubore bearing for use with SS or polished glass stirring shafts at approximately 200rpm. Consists of PTFE inner Trubore bearing, FETFE® compression saddle O-ring, glass-reinforced bushing and lock-nut with FETFE O-ring, and threaded glass adapter. The lock permits stirring in either direction without fear of unthreading. The compression saddle O-ring maintains constant force with little attention. Vacuum to 1 mm Hg. Not recommended for use with ground-glass shafts.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Trubore is a registered trademark of Ace Glass, Inc.