Documents
Z258733
Sigma-Aldrich
Ace vacuum-sublimation apparatus
Repl. CAPFE O-ring for Z258717
About This Item
UNSPSC Code:
41121800
NACRES:
NB.76
manufacturer/tradename
Ace Glass 7855881
General description
The Ace vacuum-sublimation apparatus features a greaseless O-ring sealed flange with a quick-release clamp. The top flange is ground flat, bottom mating flange has a groove for a CAPFE (fluororesin encapsulated silicone) O-ring. The complete apparatus includes an outer body, inner top, CAPFE O-ring, and quick-release clamp.